2-Cyclopentylethanol ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B22344.03 B22344.06
B22344.03EA 72.4 EUR
B22344.03 B22344.06
2-Cyclopentylethanol ≥98%
2-Cyclopentylethanol

Purity: 98+% CAS No.: 766-00-7 Molecular Formula: C7H14O Formula Weight: 114.19 Flash Point: 70°(158°F) Refractive Index: 1.4585 Boiling Point: 182° Beilstein Reference: 2069860 MDL No.: MFCD00019301

Formel: C₇H₁₄O
Siedepunkt: 182 °C
Dichte: 0,920
Flammpunkt: 70 °C (158 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00019301
CAS-Nummer: 766-00-7
EINECS: 212-156-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR