(±)-3-Methyl-1-pentanol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Methylpentan-1-ol, (±)-3-Methylpentan-1-ol, 3-Methyl-1-pentanol
B22356.06
B22356.14
B22356.22
B22356.06EA
42.4
EUR
B22356.06
B22356.14
B22356.22
(±)-3-Methyl-1-pentanol ≥98%
(±)-3-Methyl-1-pentanol
Purity: 98%
CAS No.: 589-35-5
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 58°(136°F)
Refractive Index: 1.4180
Boiling Point: 151-152°
Beilstein Reference: 1718979
UN No.: 1987
MDL No.: MFCD00002937
Formel:
C₆H₁₄O Molecular Weight: 102.18 g/mol Siedepunkt: 151…152 °C Schmelzpunkt: –70 °C Dichte: 0.823 Flammpunkt: 58 °C (136 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002937 CAS-Nummer: 589-35-5 EINECS: 209-644-9 UN: 1987 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...