N-Cbz-D-Alaninol 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
(2R)-N-Benzyloxycarbonylamino-1-propanol, N-Cbz-D-Alaninol, Z-D-Alaninol, (R)-2-(Cbz-amino)-1-propanol
B22372.03
B22372.06
B22372.14
B22372.03EA
20.3
EUR
B22372.03
B22372.06
B22372.14
N-Cbz-D-Alaninol 98%
N-Cbz-D-Alaninol
Purity: 98%
CAS No.: 61425-27-2
Molecular Formula: C11H15NO3
Formula Weight: 209.25
Storage Temperature: 2°C to 8°C
Melting Point: 82-84°
MDL No.: MFCD00672531
Formel:
C₁₁H₁₅NO₃ Molecular Weight: 209.2 g/mol Schmelzpunkt: 82…84 °C |
MDL:
MFCD00672531 CAS-Nummer: 61425-27-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...