Methyl-4-(chlormethyl)benzoat ≥97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Methyl 4-(chloromethyl)benzoate
B22476.04
B22476.04EA
39.1
EUR
B22476.04
Methyl-4-(chlormethyl)benzoat ≥97%
Methyl-4-(chlormethyl)benzoat
Purity: 97%
CAS No.: 34040-64-7
Molecular Formula: C9H9ClO2
Formula Weight: 184.62
Melting Point: 39-40°
Boiling Point: 160-165°/30mm
UN No.: 3261
MDL No.: MFCD00511224
Formel:
C₉H₉ClO₂ Molecular Weight: 184,62 g/mol Siedepunkt: 160…165 °C (30 mmHg) Schmelzpunkt: 39…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00511224 CAS-Nummer: 34040-64-7 EINECS: 251-806-6 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...