4-Cyan-4'-pentyloxybiphenyl ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Amyloxy-4'-cyanobiphenyl, 4-Cyano-4'-n-pentyloxybiphenyl, 5OCB, 4'-Amyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-[1,1'-biphenyl]-4-carbonitrile

B22477.03 B22477.06
B22477.03EA 52.3 EUR
B22477.03 B22477.06
4-Cyan-4'-pentyloxybiphenyl ≥99%
4-Cyan-4'-pentyloxybiphenyl

Purity: 99% CAS No.: 52364-71-3 Molecular Formula: C18H19NO Formula Weight: 281.42 Flash Point: 110°(230°F) Melting Point: 48° cloudy UN No.: 3439 MDL No.: MFCD00074879

Formel: C₁₈H₁₉NO
Molecular Weight: 265.35 g/mol
Schmelzpunkt: 48 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00074879
CAS-Nummer: 52364-71-3
EINECS: 257-875-9
ADR: 6.1 III

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR