Phosphor(V)bromid 95%

Lieferant: Thermo Fisher Scientific
Gefahr

B22481.14 B22481.22 B22481.36
B22481.14EA 83.8 EUR
B22481.14 B22481.22 B22481.36
Phosphor(V)bromid 95%
Phosphor(V)bromid

Purity: 95% CAS No.: 7789-69-7 Molecular Formula: PBr5 Formula Weight: 430.50 Odour: Acrid Melting Point: ca 105° dec. Boiling Point: 105-107° dec. Beilstein Reference: 3903332 Merck Reference: 14,7350 UN No.: 2691 MDL No.: MFCD00011437

Formel: PBr₅
Siedepunkt: 105…107 °C (Zers.)
Dichte: 3,61 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011437
CAS-Nummer: 7789-69-7
EINECS: 232-186-6
UN: 2691
ADR: 8,II
Merck Index: 14,7350

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR