1-Acetylpyren ≥97%
Lieferant: Thermo Scientific
Synonyme:
1-(Pyren-1-yl)ethanone
B22483.88
B22483.88EA
55.7
EUR
B22483.88
1-Acetylpyren ≥97%
1-Acetylpyren
Purity: 97%
CAS No.: 3264-21-9
Molecular Formula: C18H12O
Formula Weight: 244.29
Melting Point: 86-89°
MDL No.: MFCD00021663
Formel:
C₁₈H₁₂O Molecular Weight: 244,29 g/mol Schmelzpunkt: 86…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00021663 CAS-Nummer: 3264-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...