Sodium-1-naphthalenesulphonate, (max. 2% H₂O) 99% (Trockengewicht)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Naphthalenesulphonic acid sodium salt, Naphthalene-1-sulphonic acid sodium salt
B22488.06
B22488.14
B22488.22
B22488.06EA
28.2
EUR
B22488.06
B22488.14
B22488.22
Sodium-1-naphthalenesulphonate, (max. 2% H₂O) 99% (Trockengewicht)
Sodium-1-naphthalenesulphonate
Purity: 99%
CAS No.: 130-14-3
Molecular Formula: C10H7NaO3S
Formula Weight: 230.22
Melting Point: 299-301°
Beilstein Reference: 3639038
MDL No.: MFCD00064964
Formel:
C₁₀H₈O₃S Molecular Weight: 230.21 g/mol Schmelzpunkt: 299…301 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064964 CAS-Nummer: 130-14-3 EINECS: 204-976-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...