N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-β-alanin 99%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Boc-beta-alanine, Boc-β-Ala-OH, BOC-beta-alanine
B22522.09
B22522.88
B22522.09EA
68.9
EUR
B22522.09
B22522.88
N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-β-alanin 99%
Boc-b-Alanine
Purity: 99%
CAS No.: 3303-84-2
Molecular Formula: C8H15NO4
Formula Weight: 189.21
Storage Temperature: 2°C to 8°C
Melting Point: 73-76°
MDL No.: MFCD00037291
Formel:
C₈H₁₅NO₄ Schmelzpunkt: 73…76 °C |
MDL:
MFCD00037291 CAS-Nummer: 3303-84-2 EINECS: 221-979-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...