5-tert-Butyl-N-carbobenzoxy-L-glutamate 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Z-Glu(OtBu)-OH, N-Carbobenzoxy-L-glutamic Acid 5-tert-Butyl Ester, N-Cbz-L-glutamic Acid 5-tert-Butyl Ester, 5-tert-Butyl N-Cbz-L-glutamate
B22557.06
B22557.06EA
55.1
EUR
B22557.06
5-tert-Butyl-N-carbobenzoxy-L-glutamate 97%
5-tert-Butyl-N-carbobenzoxy-L-glutamate
Purity: 97%
CAS No.: 3886-08-6
Molecular Formula: C17H23NO6
Formula Weight: 337.37
Melting Point: 85-88°
Optical Rotation: -13° (c=2 in methanol)
MDL No.: MFCD00038274
Formel:
C₁₇H₂₃NO₆ Molecular Weight: 337,37 g/mol Schmelzpunkt: 85…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038274 CAS-Nummer: 3886-08-6 EINECS: 223-421-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...