O-tert-Butyl-L-tyrosine 99%
Lieferant: Thermo Scientific
Synonyme:
H-Tyr(tBu)-OH
B22575.03
B22575.03EA
75.3
EUR
B22575.03
O-tert-Butyl-L-tyrosine 99%
O-tert-Butyl-L-tyrosine
Purity: 99%
CAS No.: 18822-59-8
Molecular Formula: C13H19NO3
Formula Weight: 237.3
Melting Point: ca 205° dec.
MDL No.: MFCD00037202
Formel:
C₁₃H₁₉NO₃ Molecular Weight: 237,30 g/mol Schmelzpunkt: ∼ 205 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037202 CAS-Nummer: 18822-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...