5-Methoxy-2-methylbenzothiazol ≥97%

Lieferant: Thermo Scientific

B22584.06
B22584.06EA 119 EUR
B22584.06
5-Methoxy-2-methylbenzothiazol ≥97%
5-Methoxy-2-methylbenzothiazole

Purity: 97% CAS No.: 2941-69-7 Molecular Formula: C9H9NOS Formula Weight: 179.24 Refractive Index: 1.6100 Melting Point: 34-38° Beilstein Reference: 124232 MDL No.: MFCD00082664

Formel: C₉H₉NOS
Molecular Weight: 179,24 g/mol
Schmelzpunkt: 34…38 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082664
CAS-Nummer: 2941-69-7
EINECS: 220-929-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR