2-Fluor-α-methyl-4-biphenylessigsäure ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys
B22603.03
B22603.06
B22603.14
B22603.03EA
70.8
EUR
B22603.03
B22603.06
B22603.14
2-Fluor-α-methyl-4-biphenylessigsäure ≥99%
2-Fluor-α-methyl-4-biphenylessigsäure
Purity: 99%
CAS No.: 5104-49-4
Molecular Formula: C15H13FO2
Formula Weight: 244.27
Odour: Odourless
Melting Point: 112-114°
UN No.: 2811
MDL No.: MFCD00079303
Formel:
C₁₅H₁₃FO₂ Schmelzpunkt: 112…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00079303 CAS-Nummer: 5104-49-4 EINECS: 225-827-6 ADR: 6.1,III Merck Index: 14,4199 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...