2-Fluor-α-methyl-4-biphenylessigsäure ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Ansaid, 2-(2-Fluoro-[1,1'-biphenyl]-4-yl)propanoic acid, Flurbiprofen, Froben, Antadys

B22603.03 B22603.06 B22603.14
B22603.03EA 70.8 EUR
B22603.03 B22603.06 B22603.14
2-Fluor-α-methyl-4-biphenylessigsäure ≥99%
2-Fluor-α-methyl-4-biphenylessigsäure

Purity: 99% CAS No.: 5104-49-4 Molecular Formula: C15H13FO2 Formula Weight: 244.27 Odour: Odourless Melting Point: 112-114° UN No.: 2811 MDL No.: MFCD00079303

Formel: C₁₅H₁₃FO₂
Schmelzpunkt: 112…114 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00079303
CAS-Nummer: 5104-49-4
EINECS: 225-827-6
ADR: 6.1,III
Merck Index: 14,4199

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR