bisbenzimide H-33342 trihydrochloride (Hoechst 33342) 98 HPLC, TLC, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Hoechst 33342, 2′-(4-Ethoxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5′-bi-1H-benzimidazole trihydrochloride, HOE 33342, 2'-(4-Ethoxyphenyl)-5-(4-methylpiperazin-1-yl)-1H,1'H-2,5'-bibenzo[d]imidazole, bisBenzimide

MQ300

B2261-25MG B2261-100MG B2261-1G B2261-500MG
B2261-25MGEA 43.7 EUR
B2261-25MG B2261-100MG B2261-1G B2261-500MG
bisbenzimide H-33342 trihydrochloride (Hoechst 33342) 98 HPLC, TLC, Sigma-Aldrich®
bisbenzimide H-33342 trihydrochloride (Hoechst 33342)

Useful for staining DNA, chromosomes and nuclei. May be used for fluorescence microscopy or flow cytometry.
Excitation max. = 346 nm
Emission max. = 460 nm
bisBenzimide H 33342 trihydrochloride has been used for the staining of viable and apoptotic.

Formel: C₂₇H₂₈N₆O·3HCl
Molecular Weight: 561.93 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00012679
CAS-Nummer: 23491-52-3
EINECS: 200-835-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df35663a377a6af410d46bc75cb7d9eef5891fb4&vid=dadb7d35686760b2287f427c5a061eb322aa996e&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (Form) Powder
Appearance (Colour) Yellow
Solubility (Solvent) Water
Solubility (Conc) 49.00 - 51.00 mg/ml
Solubility (Turbidity) Clear to Very Slightly Hazy
Solubility (Colour) Yellow to Brown
NMR (Solvent) D2O
Identity by NMR Consistent
Purity (TLC) ≥98.00%
Purity (HPLC) ≥98.00%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR