Dibenzyldiisopropylphosphoramidit 94%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 95%
CAS No.: 108549-23-1
Molecular Formula: C20H28NO2P
Formula Weight: 345.43
Storage Temperature: 2°C to 8°C
Flash Point: 69°(156°F)
Physical Form: Liquid
Density: 1.030
Sensitivity: Moisture Sensitive
Boiling Point: 130°/0.55mm
MDL No.: MFCD00191988
Formel:
C₂₀H₂₈NO₂P Siedepunkt: 130 °C (0,55 mmHg) Dichte: 1,030 Flammpunkt: 69 °C (156 °F) |
MDL:
MFCD00191988 CAS-Nummer: 108549-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...