2-Mercaptothiophen 97% (als monomer) contains dimer

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Thiophene-2-thiol

B22642.03 B22642.06 B22642.14
B22642.03EA 30.1 EUR
B22642.03 B22642.06 B22642.14
2-Mercaptothiophen 97% (als monomer) contains dimer
2-Mercaptothiophene

Purity: 97% CAS No.: 7774-74-5 Molecular Formula: C4H4S2 Formula Weight: 116.20 Storage Temperature: 2°C to 8°C Refractive Index: 1.6170 Odour: Unpleasant Boiling Point: 62-65°/12mm Beilstein Reference: 104650 MDL No.: MFCD00051666

Formel: C₄H₄S₂
Molecular Weight: 116.20 g/mol
Siedepunkt: 62…65 °C (12 mmHg)
Dichte: 1.247
Flammpunkt: 66 °C (151 °F)
MDL: MFCD00051666
CAS-Nummer: 7774-74-5
EINECS: 231-881-1
UN: 3334
ADR: 9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR