2-Mercaptothiophen 97% (als monomer) contains dimer
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Thiophene-2-thiol
B22642.03
B22642.06
B22642.14
B22642.03EA
30.1
EUR
B22642.03
B22642.06
B22642.14
2-Mercaptothiophen 97% (als monomer) contains dimer
2-Mercaptothiophene
Purity: 97%
CAS No.: 7774-74-5
Molecular Formula: C4H4S2
Formula Weight: 116.20
Storage Temperature: 2°C to 8°C
Refractive Index: 1.6170
Odour: Unpleasant
Boiling Point: 62-65°/12mm
Beilstein Reference: 104650
MDL No.: MFCD00051666
Formel:
C₄H₄S₂ Molecular Weight: 116.20 g/mol Siedepunkt: 62…65 °C (12 mmHg) Dichte: 1.247 Flammpunkt: 66 °C (151 °F) |
MDL:
MFCD00051666 CAS-Nummer: 7774-74-5 EINECS: 231-881-1 UN: 3334 ADR: 9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...