Benzoinisopropylether ≥98%
Lieferant: Thermo Scientific
Synonyme:
2-Isopropoxy-1,2-diphenylethanone
B22654.22
B22654.22EA
106
EUR
B22654.22
Benzoinisopropylether ≥98%
Benzoinisopropylether
Purity: 98+%
CAS No.: 6652-28-4
Molecular Formula: C17H18O2
Formula Weight: 254.33
Melting Point: 76-79°
Boiling Point: 180°/10mm
MDL No.: MFCD00015025
Formel:
C₁₇H₁₈O₂ Molecular Weight: 254,33 g/mol Siedepunkt: 180 °C (10 mmHg) Schmelzpunkt: 76…79 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00015025 CAS-Nummer: 6652-28-4 EINECS: 229-677-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...