Ferulasäuremethylester ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 4-hydroxy-3-methoxycinnamate, Methyl 3-(4-hydroxy-3-methoxyphenyl)acrylate
B22657.04
B22657.09
B22657.04EA
59.8
EUR
B22657.04
B22657.09
Ferulasäuremethylester ≥99%
Ferulasäuremethylester
Purity: 99%
CAS No.: 2309-07-1
Molecular Formula: C11H12O4
Formula Weight: 208.22
Melting Point: 62-65°
MDL No.: MFCD00017208
Formel:
C₁₁H₁₂O₄ Schmelzpunkt: 62…65 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017208 CAS-Nummer: 2309-07-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...