L-Tyrosin-Methylester -hydrochlorid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
L-Methyltyrosinate hydrochloride, H-Tyr-OMe.HCl, H-Tyr-OMe·HCl
B22683.14
B22683.22
B22683.14EA
51.3
EUR
B22683.14
B22683.22
L-Tyrosin-Methylester -hydrochlorid ≥98%
L-Tyrosin-Methylester -hydrochlorid
Purity: 98+%
CAS No.: 3417-91-2
Molecular Formula: C10H13NO3.HCl
Formula Weight: 231.68
Storage Temperature: 2°C to 8°C
Melting Point: 187° dec.
Beilstein Reference: 3917353
Optical Rotation: -5.2° (c= 2.4 in water)
MDL No.: MFCD00012607
Formel:
C₁₀H₁₃NO₃ Molecular Weight: 231.68 g/mol Schmelzpunkt: 187 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00012607 CAS-Nummer: 3417-91-2 EINECS: 222-313-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...