2-Iod-4-(trifluormethyl)anilin ≥99%

Lieferant: Thermo Fisher Scientific
Gefahr

B22691.09 B22691.18 B22691.30 AB131379.0250
B22691.09EA 64 EUR
B22691.09 B22691.18 B22691.30 ABCRAB131379.0250
2-Iod-4-(trifluormethyl)anilin ≥99%
2-Iod-4-(trifluormethyl)anilin

Purity: 99% CAS No.: 163444-17-5 Molecular Formula: C7H5F3IN Formula Weight: 287.02 Melting Point: 49-52° MDL No.: MFCD02683097

Formel: C₇H₅F₃IN
Schmelzpunkt: 49…52 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02683097
CAS-Nummer: 163444-17-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR