2-Iod-4-(trifluormethyl)anilin ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
B22691.09
B22691.18
B22691.30
AB131379.0250
B22691.09EA
64
EUR
B22691.09
B22691.18
B22691.30
ABCRAB131379.0250
2-Iod-4-(trifluormethyl)anilin ≥99%
2-Iod-4-(trifluormethyl)anilin
Purity: 99%
CAS No.: 163444-17-5
Molecular Formula: C7H5F3IN
Formula Weight: 287.02
Melting Point: 49-52°
MDL No.: MFCD02683097
Formel:
C₇H₅F₃IN Schmelzpunkt: 49…52 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683097 CAS-Nummer: 163444-17-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...