4-Piperidinol ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 5382-16-1
Molecular Formula: C5H11NO
Formula Weight: 101.15
Flash Point: 107°(224°F)
Odour: Amine-like
pH: 11.5-11.7
Melting Point: 86-90°
Boiling Point: 108-114°/10mm
Beilstein Reference: 102738
UN No.: 3263
MDL No.: MFCD00005999
Formel:
C₅H₁₁NO Siedepunkt: 108…114 °C (10 mmHg) Schmelzpunkt: 86…90 °C Flammpunkt: 107 °C (224 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005999 CAS-Nummer: 5382-16-1 EINECS: 226-373-1 UN: 3263 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...