(Furfurylthio)aceton 96%

Lieferant: Thermo Fisher Scientific
Achtung

B22724.06
B22724.06EA 38.5 EUR
B22724.06
(Furfurylthio)aceton 96%
(Furfurylthio)aceton

Purity: 96% CAS No.: 58066-86-7 Molecular Formula: C8H10O2S Formula Weight: 170.23 MDL No.: MFCD02683098

Formel: C₈H₁₀O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD02683098
CAS-Nummer: 58066-86-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR