2-Methylnaphtho[1,2-d]thiazole 98%

Lieferant: Thermo Fisher Scientific

B22725.06
B22725.06EA 55.1 EUR
B22725.06
2-Methylnaphtho[1,2-d]thiazole 98%
2-Methylnaphtho[1,2-d]thiazole

Purity: 98% CAS No.: 2682-45-3 Molecular Formula: C12H9NS Formula Weight: 199.27 Melting Point: 95-97° MDL No.: MFCD00004951

Formel: C₁₂H₉NS
Schmelzpunkt: 95…97 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004951
CAS-Nummer: 2682-45-3
EINECS: 220-240-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR