3,4-Dimethylanisol ≥99%

Lieferant: Thermo Scientific
Achtung

B22735.18
B22735.18EA 70.1 EUR
B22735.18
3,4-Dimethylanisol ≥99%
3,4-Dimethylanisol

Purity: 99% CAS No.: 4685-47-6 Molecular Formula: C9H12O Formula Weight: 136.19 Flash Point: 75°(167°F) Refractive Index: 1.5175 Boiling Point: 200-201° Beilstein Reference: 2040907 MDL No.: MFCD00008396

Formel: C₉H₁₂O
Molecular Weight: 136,19 g/mol
Siedepunkt: 200…201 °C
Dichte: 0,975
Flammpunkt: 75 °C (167 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008396
CAS-Nummer: 4685-47-6
EINECS: 225-142-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR