2-Benzo[b]thiophenboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B22835.03
B22835.06
B22835.14
B22835.03EA
34.1
EUR
B22835.03
B22835.06
B22835.14
2-Benzo[b]thiophenboronsäure ≥98%
2-Benzo[b]thiophenboronsäure
Purity: 98%
CAS No.: 98437-23-1
Molecular Formula: C8H7BO2S
Formula Weight: 178.02
Melting Point: ca 255°
Beilstein Reference: 1637924
Formel:
C₈H₇BO₂S Schmelzpunkt: ∼ 255 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075674 CAS-Nummer: 98437-23-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...