3-Brom-1-propanol ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Brom-3-hydroxypropan, Trimethylenbromhydrin
B22857.06
B22857.14
B22857.22
B22857.06EA
35.6
EUR
B22857.06
B22857.14
B22857.22
3-Brom-1-propanol ≥97%
3-Brom-1-propanol
Purity: 97+%
CAS No.: 627-18-9
Molecular Formula: C3H7BrO
Formula Weight: 138.99
Storage Temperature: 2°C to 8°C
Flash Point: 93°(199°F)
Refractive Index: 1.4885
Boiling Point: 71-73°/11mm
Beilstein Reference: 969160
MDL No.: MFCD00002942
Formel:
BrH₂CCH₂CH₂OH Siedepunkt: 71…73 °C (11 mmHg) Dichte: 1,590 Flammpunkt: 93 °C (199 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00002942 CAS-Nummer: 627-18-9 EINECS: 210-986-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...