1,12-Dodecandiol ≥99%
Lieferant: Thermo Scientific
Synonyme:
1,12-Dihydroxydodecan, Dodecamethylenglycol
B22885.14
B22885.14EA
41.9
EUR
B22885.14
1,12-Dodecandiol ≥99%
1,12-Dodecandiol
Purity: 99%
CAS No.: 5675-51-4
Molecular Formula: HO(CH2)12OH
Formula Weight: 202.34
Melting Point: 81-84°
Boiling Point: 188-190°/12mm
MDL No.: MFCD00004755
Formel:
C₁₂H₂₆O₂ Molecular Weight: 202,34 g/mol Siedepunkt: 188…190 °C (12 mmHg) Schmelzpunkt: 81…84 °C Dichte: 1,08 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004755 CAS-Nummer: 5675-51-4 EINECS: 227-133-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...