4-(Trifluormethylsulfonyl)benzonitril ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

B22907.03 B22907.06
B22907.03EA 42.5 EUR
B22907.03 B22907.06
4-(Trifluormethylsulfonyl)benzonitril ≥97%
4-(Trifluormethylsulfonyl)benzonitril

Purity: 97+% CAS No.: 312-21-0 Molecular Formula: C8H4F3NO2S Formula Weight: 235.18 Melting Point: 87-92° UN No.: 3439 MDL No.: MFCD01631631

Formel: C₈H₄F₃NO₂S
Schmelzpunkt: 87…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01631631
CAS-Nummer: 312-21-0
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR