2-Amino-4-(2-methyl-2-propanyl)phenol 97%

Lieferant: Thermo Fisher Scientific
Gefahr

B22964.06 B22964.14
B22964.06EA 31 EUR
B22964.06 B22964.14
2-Amino-4-(2-methyl-2-propanyl)phenol 97%
2-Amino-4-tert-butylphenol

Purity: 97% CAS No.: 1199-46-8 Molecular Formula: C10H15NO Formula Weight: 165.23 Melting Point: 162-164° MDL No.: MFCD00007698

Formel: C₁₀H₁₅NO
Schmelzpunkt: 162…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007698
CAS-Nummer: 1199-46-8
EINECS: 214-844-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR