N,N'-Ethylendianilin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B23098.14 B23098.06 B23098.22
B23098.14EA 74.6 EUR
B23098.14 B23098.06 B23098.22
N,N'-Ethylendianilin ≥97%
N,N'-Ethylendianilin

Purity: 98+% CAS No.: 150-61-8 Molecular Formula: C6H5NHCH2CH2NHC6H5 Formula Weight: 212.30 Melting Point: 64-68° Boiling Point: 228°/12mm Merck Reference: 14,2990 MDL No.: MFCD00003019

Formel: C₆H₅NHCH₂CH₂NHC₆H₅
Siedepunkt: 228 °C (12 mmHg)
Schmelzpunkt: 64…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003019
CAS-Nummer: 150-61-8
EINECS: 205-765-6
Merck Index: 14,2990

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR