4-Methoxybenzophenon ≥98%
Lieferant: Thermo Scientific
Achtung
|
Synonyme:
4-Benzoylanisol, p-Methoxybenzophenon
B23123.14
B23123.14EA
48.7
EUR
B23123.14
4-Methoxybenzophenon ≥98%
4-Methoxybenzophenon
Purity: 98+%
CAS No.: 611-94-9
Molecular Formula: C14H12O2
Formula Weight: 212.25
Melting Point: 60-63°
Boiling Point: 354-356°
MDL No.: MFCD00008403
Formel:
C₁₄H₁₂O₂ Molecular Weight: 212,25 g/mol Siedepunkt: 354…356 °C Schmelzpunkt: 60…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008403 CAS-Nummer: 611-94-9 EINECS: 210-285-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...