5-Methyl-2-thiophenboronsäure ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B23138.03
B23138.06
B23138.03EA
42.5
EUR
B23138.03
B23138.06
5-Methyl-2-thiophenboronsäure ≥98%
5-Methyl-2-thiopheneboronic acid
Purity: 98%
CAS No.: 162607-20-7
Molecular Formula: C5H7BO2S
Formula Weight: 142.01
Storage Temperature: 2°C to 8°C
Melting Point: ca 160° dec.
Beilstein Reference: 7019569
MDL No.: MFCD01318166
Formel:
C₅H₇BO₂S Schmelzpunkt: ∼ 160 °C (Zersetzung) |
MDL:
MFCD01318166 CAS-Nummer: 162607-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...