trans-2,5-Dimethylpiperazin ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

B23150.06 B23150.14 B23150.22
B23150.06EA 35.2 EUR
B23150.06 B23150.14 B23150.22
trans-2,5-Dimethylpiperazin ≥98%
trans-2,5-Dimethylpiperazin

Purity: 98% CAS No.: 2815-34-1 Molecular Formula: C6H14N2 Formula Weight: 114.19 Flash Point: 58°(136°F) Melting Point: 115-119° Boiling Point: 162-165° Beilstein Reference: 79886 UN No.: 2925 MDL No.: MFCD00070516

Formel: C₆H₁₄N₂
Molecular Weight: 114.19 g/mol
Siedepunkt: 162…165 °C
Schmelzpunkt: 115…119 °C
Flammpunkt: 58 °C (136 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00070516
CAS-Nummer: 2815-34-1
ADR: 4.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR