Dipropylsulfid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

B23155.14 B23155.22 B23155.36
B23155.14EA 64.9 EUR
B23155.14 B23155.22 B23155.36
Dipropylsulfid ≥98%
Dipropylsulfid

Purity: 98+% CAS No.: 111-47-7 Molecular Formula: (CH3CH2CH2)2S Formula Weight: 118.24 Flash Point: 28°(82°F) Refractive Index: 1.4487 Odour: Stench Melting Point: -102° Boiling Point: 141-142° Beilstein Reference: 1719002 Merck Reference: 14,7868 UN No.: 1993 MDL No.: MFCD00009379

Formel: C₆H₁₄S
Siedepunkt: 141…142 °C
Schmelzpunkt: –102 °C
Dichte: 0,838
Flammpunkt: 28 °C (82 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009379
CAS-Nummer: 111-47-7
EINECS: 203-873-8
UN: 1993
ADR: 3,III
Merck Index: 14,7868

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR