Dipropylsulfid ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
B23155.14
B23155.22
B23155.36
B23155.14EA
64.9
EUR
B23155.14
B23155.22
B23155.36
Dipropylsulfid ≥98%
Dipropylsulfid
Purity: 98+%
CAS No.: 111-47-7
Molecular Formula: (CH3CH2CH2)2S
Formula Weight: 118.24
Flash Point: 28°(82°F)
Refractive Index: 1.4487
Odour: Stench
Melting Point: -102°
Boiling Point: 141-142°
Beilstein Reference: 1719002
Merck Reference: 14,7868
UN No.: 1993
MDL No.: MFCD00009379
Formel:
C₆H₁₄S Siedepunkt: 141…142 °C Schmelzpunkt: –102 °C Dichte: 0,838 Flammpunkt: 28 °C (82 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009379 CAS-Nummer: 111-47-7 EINECS: 203-873-8 UN: 1993 ADR: 3,III Merck Index: 14,7868 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...