4-Methyl-5-nonanol (Gemisch der Stereoisomere) ≥98%
Lieferant: Thermo Fisher Scientific
B23167.09
B23167.18
B23167.30
B23167.09EA
49.7
EUR
B23167.09
B23167.18
B23167.30
4-Methyl-5-nonanol (Gemisch der Stereoisomere) ≥98%
4-Methyl-5-nonanol (Gemisch der Stereoisomere)
Purity: 98%
CAS No.: 154170-44-2
Molecular Formula: C10H22O
Formula Weight: 158.29
Refractive Index: 1.4360
Formel:
C₁₀H₂₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00046696 CAS-Nummer: 154170-44-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...