Tetraethylammoniumfluorid Hydrat ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

B23173.06 B23173.14 B23173.22
B23173.06EA 62.9 EUR
B23173.06 B23173.14 B23173.22
Tetraethylammoniumfluorid Hydrat ≥97%
Tetraethylammoniumfluorid Hydrat

Purity: 97% CAS No.: 98330-04-2 Molecular Formula: (CH3CH2)4NF.xH2O Formula Weight: 149.26(anhy) Beilstein Reference: 4152659 MDL No.: MFCD00149990

Formel: C₈H₂₀FN·xH₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00149990
CAS-Nummer: 98330-04-2
EINECS: 211-559-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR