(±)-2-Phenylbuttersäure ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: (±)-2-Phenylbutansäure

B23222.22
B23222.22EA 53.6 EUR
B23222.22
(±)-2-Phenylbuttersäure ≥98%
(±)-2-Phenylbuttersäure

Purity: 98% CAS No.: 90-27-7 Molecular Formula: C10H12O2 Formula Weight: 164.20 Flash Point: 110°(230°F) Refractive Index: 1.5150 Melting Point: 40-44° Boiling Point: 270-272° Beilstein Reference: 509876 MDL No.: MFCD00002667

Formel: C₁₀H₁₂O₂
Molecular Weight: 164,2 g/mol
Siedepunkt: 270…272 °C
Schmelzpunkt: 40…44 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00002667
CAS-Nummer: 90-27-7
EINECS: 201-982-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR