(3-Brompropyl)ammoniumbromid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Amino-3-brompropan Hydrobromid, 3-Brompropylamin Hydrobromid

B23254.14 B23254.22
B23254.14EA 47.8 EUR
B23254.14 B23254.22
(3-Brompropyl)ammoniumbromid 98%
3-Brompropylamine hydrobromide

Purity: 98% CAS No.: 5003-71-4 Molecular Formula: Br(CH2)3NH2.HBr Formula Weight: 218.93 Melting Point: 172-174° Beilstein Reference: 3906418

Formel: BrH₂CCH₂CH₂NH₃Br
Molecular Weight: 218.92 g/mol
Schmelzpunkt: 172…174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012912
CAS-Nummer: 5003-71-4
EINECS: 225-675-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR