Methylisonicotinat ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Pyridinecarboxylic acid methyl ester, 4-Picolinic acid methyl ester, Methyl 4-pyridinecarboxylate, Pyridine-4-carboxylic acid methyl ester, Isonicotinic acid methyl ester
B23263.14
B23263.22
B23263.36
AB132158.0500
B23263.14EA
30.5
EUR
B23263.14
B23263.22
B23263.36
ABCRAB132158.0500
Methylisonicotinat ≥98%
Methylisonicotinat
Purity: 98%
CAS No.: 2459-09-8
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Flash Point: 82°(180°F)
Refractive Index: 1.5135
Odour: Characteristic
Melting Point: 8-8.5°
Boiling Point: 207-209°
MDL No.: MFCD00006427
Formel:
C₇H₇NO₂ Molecular Weight: 137.14 g/mol Siedepunkt: 207…209 °C Schmelzpunkt: 8…8.5 °C Dichte: 1.016 Flammpunkt: 82 °C (180 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00006427 CAS-Nummer: 2459-09-8 EINECS: 219-456-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...