1-Iod-3,3,3-trifluorpropan ≥98%
Lieferant: Thermo Fisher Scientific
B23284.03
B23284.06
B23284.03EA
26.8
EUR
B23284.03
B23284.06
1-Iod-3,3,3-trifluorpropan ≥98%
1-Iod-3,3,3-trifluorpropan
Purity: 98+%
CAS No.: 460-37-7
Molecular Formula: CF3CH2CH2I
Formula Weight: 223.96
Flash Point: None
Refractive Index: 1.4200
Boiling Point: 89-90°
Beilstein Reference: 1698182
MDL No.: MFCD00038531
Formel:
C₃H₄F₃I Siedepunkt: 89…90 °C Dichte: 1,911 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038531 CAS-Nummer: 460-37-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...