1-Chlor-3-nitrobenzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Chlornitrobenzol, MNCB
B23423.18
B23423.30
B23423.18EA
28.9
EUR
B23423.18
B23423.30
1-Chlor-3-nitrobenzol ≥98%
1-Chlor-3-nitrobenzol
Purity: 98%
CAS No.: 121-73-3
Molecular Formula: C6H4ClNO2
Formula Weight: 157.56
Flash Point: 124°(255°F)
Melting Point: 43-47°
Boiling Point: 235-236°
Beilstein Reference: 1907947
Merck Reference: 14,2151
UN No.: 1578
MDL No.: MFCD00007202
Formel:
C₆H₄ClNO₂ Molecular Weight: 157.56 g/mol Siedepunkt: 235…236 °C Schmelzpunkt: 43…47 °C Dichte: 1.34 Flammpunkt: 124 °C (255 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007202 CAS-Nummer: 121-73-3 EINECS: 204-496-1 UN: 1578 ADR: 6.1 II Merck Index: 13,02170 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...