Patton-Reeder-Reagenz (Calconcarbonsäure), (max. 10% H₂O)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-3-naphthincarbonsäure, Cal-Red®, 2,2'-Dihydroxy-4'-sulfo-1,1'-azonaphthalin-3-carbonsäure, 3-Hydroxy-4-(2-hydroxy-4-sulfo-1-naphthylazo)naphthalin-2-carbonsäure
B23459.09
B23459.18
B23459.09EA
59.9
EUR
B23459.09
B23459.18
Patton-Reeder-Reagenz (Calconcarbonsäure), (max. 10% H₂O)
Patton-Reeder-Reagenz (Calconcarbonsäure)
CAS No.: 3737-95-9
Molecular Formula: C21H14N2O7S
Formula Weight: 438.42
Melting Point: ca 300° dec.
Beilstein Reference: 727492
MDL No.: MFCD00004078
Formel:
C₂₁H₁₄N₂O₇S Molecular Weight: 438.42 g/mol Schmelzpunkt: ∼ 300 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004078 CAS-Nummer: 3737-95-9 EINECS: 223-117-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...