Patton-Reeder-Reagenz (Calconcarbonsäure), (max. 10% H₂O)

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Hydroxy-1-(2-hydroxy-4-sulfo-1-naphthylazo)-3-naphthincarbonsäure, Cal-Red®, 2,2'-Dihydroxy-4'-sulfo-1,1'-azonaphthalin-3-carbonsäure, 3-Hydroxy-4-(2-hydroxy-4-sulfo-1-naphthylazo)naphthalin-2-carbonsäure

B23459.09 B23459.18
B23459.09EA 59.9 EUR
B23459.09 B23459.18
Patton-Reeder-Reagenz (Calconcarbonsäure), (max. 10% H₂O)
Patton-Reeder-Reagenz (Calconcarbonsäure)

CAS No.: 3737-95-9 Molecular Formula: C21H14N2O7S Formula Weight: 438.42 Melting Point: ca 300° dec. Beilstein Reference: 727492 MDL No.: MFCD00004078

Formel: C₂₁H₁₄N₂O₇S
Molecular Weight: 438.42 g/mol
Schmelzpunkt: ∼ 300 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004078
CAS-Nummer: 3737-95-9
EINECS: 223-117-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR