3-Chlor-4-[(trifluormethyl)thio]anilin ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
B23500.03
B23500.03EA
70.7
EUR
B23500.03
3-Chlor-4-[(trifluormethyl)thio]anilin ≥97%
3-Chlor-4-[(trifluormethyl)thio]anilin
Purity: 97%
CAS No.: 64628-74-6
Molecular Formula: C7H5ClF3NS
Formula Weight: 227.64
Melting Point: 41-44°
Beilstein Reference: 6314200
UN No.: 2811
MDL No.: MFCD00190131
Formel:
C₇H₅ClF₃NS Schmelzpunkt: 41…44 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00190131 CAS-Nummer: 64628-74-6 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...