2'-Chlor-5'-(trifluormethyl)acetophenon ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

B23524.03 B23524.06
B23524.03EA 31.9 EUR
B23524.03 B23524.06
2'-Chlor-5'-(trifluormethyl)acetophenon ≥97%
2'-Chlor-5'-(trifluormethyl)acetophenon

Purity: 97+% CAS No.: 71648-45-8 Molecular Formula: C9H6ClF3O Formula Weight: 222.60 MDL No.: MFCD00045092

Formel: C₉H₆ClF₃O
Molecular Weight: 222.59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00045092
CAS-Nummer: 71648-45-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR