3-Fluorbenzylamin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
(3-Fluorophenyl)methanamine
B23527.06
B23527.09
B23527.14
B23527.06EA
64.3
EUR
B23527.06
B23527.09
B23527.14
3-Fluorbenzylamin 97%
3-Fluorbenzylamin
Purity: 97%
CAS No.: 100-82-3
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 71°(159°F)
Refractive Index: 1.5150
Odour: Amine-like
Boiling Point: 186°
Beilstein Reference: 1446928
UN No.: 2735
MDL No.: MFCD00008113
Formel:
C₇H₈FN Siedepunkt: 186 °C Dichte: 1,097 Flammpunkt: 71 °C (159 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008113 CAS-Nummer: 100-82-3 EINECS: 202-891-3 UN: 2735 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...